4'-Bromo-2'-fluoro-[1,1'-biphenyl]-4-ol ≥95%

Supplier: AOBChem USA

Warning

57556-500MG 57556-1G 57556-5G 57556-25G
77329-976EA 126.69 USD
77329-976 77344-214 77366-280 77378-156
4'-Bromo-2'-fluoro-[1,1'-biphenyl]-4-ol ≥95%
4'-Bromo-2'-fluoro-[1,1'-biphenyl]-4-ol

Precaución: for Research Use Only

Formula: C₁₂H₈BrFO
MW: 267.094 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564456
CAS Number: 116831-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR