(2-Fluoro-4'-methyl-[1,1'-biphenyl]-4-yl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

57548-500MG 57548-1G 57548-5G 57548-25G
77329-970EA 179.22 USD
77329-970 77346-722 77366-274 77388-014
(2-Fluoro-4'-methyl-[1,1'-biphenyl]-4-yl)boronic acid ≥97%
(2-Fluoro-4'-methyl-[1,1'-biphenyl]-4-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₂BFO₂
MW: 230.043 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29078681
CAS Number: 1514897-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR