5-Iodo-2-(trifluoromethoxy)aniline ≥97%

Supplier: AOBChem USA

Warning

57533-500MG 57533-1G 57533-5G 57533-25G
77329-968EA 196.73 USD
77329-968 77353-932 77366-272 77388-012
5-Iodo-2-(trifluoromethoxy)aniline ≥97%
5-Iodo-2-(trifluoromethoxy)aniline

Precaución: for Research Use Only

Formula: C₇H₅F₃INO
Storage Temperature: Ambient
MDL Number: MFCD34534558
CAS Number: 1544-90-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR