4-Iodo-2-nitro-1-(trifluoromethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

57532-500MG 57532-1G 57532-5G 57532-25G
77329-966EA 140.08 USD
77329-966 77344-756 77366-270 77388-010
4-Iodo-2-nitro-1-(trifluoromethoxy)benzene ≥97%
4-Iodo-2-nitro-1-(trifluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₇H₃F₃INO₃
Storage Temperature: Ambient
MDL Number: MFCD13194453
CAS Number: 655-71-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR