Ethyl 6-ethoxy-3-fluoro-2-(methylthio)benzoate ≥97%

Supplier: AOBChem USA

Warning

57522-500MG 57522-1G 57522-5G 57522-25G
77329-962EA 254.41 USD
77329-962 77353-926 77366-266 77388-006
Ethyl 6-ethoxy-3-fluoro-2-(methylthio)benzoate ≥97%
Ethyl 6-ethoxy-3-fluoro-2-(methylthio)benzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₅FO₃S
MW: 258.309 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758219
CAS Number: 2764729-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR