(3-Ethoxy-6-fluoro-2-iodophenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

57517-500MG 57517-1G 57517-5G
77336-138EA 378.01 USD
77336-138 77353-922 77378-150
(3-Ethoxy-6-fluoro-2-iodophenyl)(methyl)sulfane ≥95%
(3-Ethoxy-6-fluoro-2-iodophenyl)(methyl)sulphane

Precaución: for Research Use Only

Formula: C₉H₁₀FIOS
MW: 312.143 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758214
CAS Number: 2705799-53-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR