Methyl 6-ethoxy-3-fluoro-2-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

57513-500MG 57513-1G 57513-5G
77339-218EA 568.56 USD
77339-218 77353-916 77378-146
Methyl 6-ethoxy-3-fluoro-2-methylbenzoate ≥97%
Methyl 6-ethoxy-3-fluoro-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₃
MW: 212.218 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34179987
CAS Number: 2387206-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR