Ethyl 6-ethoxy-3-fluoro-2-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

57512-500MG 57512-1G 57512-5G
77339-686EA 614.91 USD
77339-686 77353-914 77378-144
Ethyl 6-ethoxy-3-fluoro-2-methylbenzoate ≥97%
Ethyl 6-ethoxy-3-fluoro-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₅FO₃
MW: 226.244 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34179988
CAS Number: 2383669-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR