6-Ethoxy-3-fluoro-2-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

57511-500MG 57511-1G 57511-5G
77338-766EA 517.06 USD
77338-766 77353-912 77378-142
6-Ethoxy-3-fluoro-2-methylbenzoic acid ≥97%
6-Ethoxy-3-fluoro-2-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₃
MW: 198.191 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564451
CAS Number: 2386653-62-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR