(4-Chloro-5-fluoro-2-(trifluoromethyl)phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

57507-500MG 57507-1G 57507-5G
77340-172EA 810.61 USD
77340-172 77353-906 77378-136
(4-Chloro-5-fluoro-2-(trifluoromethyl)phenyl)methanol ≥97%
(4-Chloro-5-fluoro-2-(trifluoromethyl)phenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₅ClF₄O
Storage Temperature: Ambient
MDL Number: MFCD34758211
CAS Number: 2384836-83-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR