2-Bromo-4-fluoro-3-iodo-1-isopropoxybenzene ≥97%

Supplier: AOBChem USA

Warning

57504-500MG 57504-1G 57504-5G 57504-25G
77329-954EA 257.5 USD
77329-954 77353-902 77366-260 77388-000
2-Bromo-4-fluoro-3-iodo-1-isopropoxybenzene ≥97%
2-Bromo-4-fluoro-3-iodo-1-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₉H₉BrFIO
Storage Temperature: Ambient
MDL Number: MFCD34564448
CAS Number: 2734773-52-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR