2-Bromo-6-methoxy-4-methylphenol ≥95%

Supplier: AOBChem USA

Warning

57499-500MG 57499-1G 57499-5G 57499-25G
77329-952EA 276.04 USD
77329-952 77353-900 77366-258 77387-998
2-Bromo-6-methoxy-4-methylphenol ≥95%
2-Bromo-6-methoxy-4-methylphenol

Precaución: for Research Use Only

Formula: C₈H₉BrO₂
Storage Temperature: Ambient
MDL Number: MFCD22414705
CAS Number: 71119-08-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR