2,3-dimethoxy-5-methylbenzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
57498-500MG
57498-1G
57498-5G
77337-496EA
415.09
USD
77337-496
77353-898
77378-132
2,3-dimethoxy-5-methylbenzoic acid ≥97%
2,3-dimethoxy-5-methylbenzoic acid
Precaución: for Research Use Only
Formula:
C₁₀H₁₂O₄ Storage Temperature: Ambient |
MDL Number:
MFCD20638928 CAS Number: 5653-56-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...