Ethyl 4-bromo-2-fluoro-3-methoxy-5-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

57490-500MG 57490-1G 57490-5G
77329-948EA 347.11 USD
77329-948 77353-896 77378-128
Ethyl 4-bromo-2-fluoro-3-methoxy-5-methylbenzoate ≥97%
Ethyl 4-bromo-2-fluoro-3-methoxy-5-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₂BrFO₃
MW: 291.114 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758205
CAS Number: 2118961-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR