(4-Bromo-2-fluoro-3-methoxy-5-methylphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

57487-500MG 57487-1G 57487-5G
77336-156EA 379.04 USD
77336-156 77353-894 77378-126
(4-Bromo-2-fluoro-3-methoxy-5-methylphenyl)methanol ≥97%
(4-Bromo-2-fluoro-3-methoxy-5-methylphenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₀BrFO₂
MW: 249.077 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758203
CAS Number: 1897432-83-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR