2,6-Dichloro-4-isopropylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

57480-500MG 57480-1G
77340-490EA 1263.81 USD
77340-490 77353-892
2,6-Dichloro-4-isopropylbenzoic acid ≥95%
2,6-Dichloro-4-isopropylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀Cl₂O₂
MW: 233.091 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758197
CAS Number: 2901096-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR