2-Iodo-4-isopropylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

57477-500MG 57477-1G 57477-5G
77338-354EA 481.01 USD
77338-354 77353-890 77378-124
2-Iodo-4-isopropylbenzaldehyde ≥97%
2-Iodo-4-isopropylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₁IO
Storage Temperature: Ambient
MDL Number: MFCD29119907
CAS Number: 1289043-21-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR