Methyl 3-methyl-5-(methylthio)benzoate ≥97%

Supplier: AOBChem USA

Warning

57474-500MG 57474-1G 57474-5G 57474-25G
77329-942EA 186.43 USD
77329-942 77353-888 77366-250 77387-992
Methyl 3-methyl-5-(methylthio)benzoate ≥97%
Methyl 3-methyl-5-(methylthio)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD22489115
CAS Number: 1822631-53-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR