(3-Fluoro-2,6-dimethoxyphenyl)(methyl)sulphane ≥97%

Supplier: AOBChem USA

Warning

57467-500MG 57467-1G 57467-5G 57467-25G
77329-938EA 103 USD
77329-938 77343-032 77366-246 77378-120
(3-Fluoro-2,6-dimethoxyphenyl)(methyl)sulphane ≥97%
(3-Fluoro-2,6-dimethoxyphenyl)(methyl)sulphane

Precaución: for Research Use Only

Formula: C₉H₁₁FO₂S
Storage Temperature: Ambient
MDL Number: MFCD28793153
CAS Number: 1806337-75-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR