(3-Iodo-5-methylphenyl)(methyl)sulphane ≥97%

Supplier: AOBChem USA

Warning

57466-500MG 57466-1G 57466-5G 57466-25G
77329-936EA 126.69 USD
77329-936 77344-152 77366-244 77378-118
(3-Iodo-5-methylphenyl)(methyl)sulphane ≥97%
(3-Iodo-5-methylphenyl)(methyl)sulphane

Precaución: for Research Use Only

Formula: C₈H₉IS
Storage Temperature: Ambient
MDL Number: MFCD22489959
CAS Number: 1824663-49-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR