2,2-Dichloro-1-(2-fluoro-3-(methylthio)phenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

57450-500MG 57450-1G 57450-5G
77338-062EA 461.44 USD
77338-062 77353-886 77378-116
2,2-Dichloro-1-(2-fluoro-3-(methylthio)phenyl)ethanol ≥95%
2,2-Dichloro-1-(2-fluoro-3-(methylthio)phenyl)ethanol

Precaución: for Research Use Only

Formula: C₉H₉Cl₂FOS
MW: 255.137 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758191
CAS Number: 2807471-26-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR