Methyl 2-fluoro-6-methoxy-4-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

57423-500MG 57423-1G 57423-5G 57423-25G
77329-934EA 170.98 USD
77329-934 77346-400 77366-242 77387-990
Methyl 2-fluoro-6-methoxy-4-methylbenzoate ≥97%
Methyl 2-fluoro-6-methoxy-4-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₃
MW: 198.191 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758188
CAS Number: 2363154-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR