Ethyl 2-fluoro-6-methoxy-4-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

57422-500MG 57422-1G 57422-5G 57422-25G
77329-932EA 170.98 USD
77329-932 77346-398 77366-240 77387-988
Ethyl 2-fluoro-6-methoxy-4-methylbenzoate ≥97%
Ethyl 2-fluoro-6-methoxy-4-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₃
Storage Temperature: Ambient
MDL Number: MFCD34758187
CAS Number: 2387217-92-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR