Methyl 4-chloro-3-ethoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

57420-500MG 57420-1G 57420-5G 57420-25G
77329-930EA 234.84 USD
77329-930 77353-884 77366-238 77387-986
Methyl 4-chloro-3-ethoxybenzoate ≥97%
Methyl 4-chloro-3-ethoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO₃
Storage Temperature: Ambient
MDL Number: MFCD19440067
CAS Number: 1379365-32-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR