1-Chloro-2-ethoxy-4-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

57419-500MG 57419-1G 57419-5G 57419-25G
77329-928EA 257.5 USD
77329-928 77353-882 77366-236 77387-984
1-Chloro-2-ethoxy-4-iodobenzene ≥97%
1-Chloro-2-ethoxy-4-iodobenzene

Precaución: for Research Use Only

Formula: C₈H₈ClIO
MW: 282.506 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32649112
CAS Number: 2741581-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR