2,5-Dimethyl-3-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

57410-500MG 57410-1G 57410-5G 57410-25G
77329-924EA 266.77 USD
77329-924 77353-878 77366-234 77387-982
2,5-Dimethyl-3-(methylthio)benzoic acid ≥97%
2,5-Dimethyl-3-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD34758181
CAS Number: 2901102-20-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR