4-Bromo-2-fluoro-3-methylbenzamide ≥95%

Supplier: AOBChem USA

Warning

57391-500MG 57391-1G 57391-25G 57391-5G
77329-922EA 290.46 USD
77329-922 77353-876 77387-980 77378-112
4-Bromo-2-fluoro-3-methylbenzamide ≥95%
4-Bromo-2-fluoro-3-methylbenzamide

Precaución: for Research Use Only

Formula: C₈H₇BrFNO
Storage Temperature: Ambient
MDL Number: MFCD32696812
CAS Number: 1228014-43-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR