2-Fluoro-4-isopropoxypyridine ≥97%

Supplier: AOBChem USA

Warning

57374-500MG 57374-1G 57374-5G 57374-25G
77329-920EA 181.28 USD
77329-920 77346-864 77366-232 77387-978
2-Fluoro-4-isopropoxypyridine ≥97%
2-Fluoro-4-isopropoxypyridine

Precaución: for Research Use Only

Formula: C₈H₁₀FNO
Storage Temperature: Ambient
MDL Number: MFCD23114426
CAS Number: 175965-84-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR