4-Ethoxy-2-fluoronicotinaldehyde ≥97%

Supplier: AOBChem USA

Warning

57371-500MG 57371-1G 57371-5G
77335-854EA 367.71 USD
77335-854 77353-874 77378-110
4-Ethoxy-2-fluoronicotinaldehyde ≥97%
4-Ethoxy-2-fluoronicotinaldehyde

Precaución: for Research Use Only

Formula: C₈H₈FNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD34564440
CAS Number: 2734777-46-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR