2-Fluoro-4-methoxy-3-(methylthio)pyridine ≥97%

Supplier: AOBChem USA

Warning

57361-500MG 57361-1G 57361-5G 57361-25G
77329-918EA 198.79 USD
77329-918 77353-872 77366-230 77387-976
2-Fluoro-4-methoxy-3-(methylthio)pyridine ≥97%
2-Fluoro-4-methoxy-3-(methylthio)pyridine

Precaución: for Research Use Only

Formula: C₇H₈FNOS
MW: 173.208 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564439
CAS Number: 2734779-44-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR