2-Fluoro-3-iodo-4-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

57360-500MG 57360-1G 57360-5G
77337-920EA 451.14 USD
77337-920 77353-870 77378-108
2-Fluoro-3-iodo-4-methoxypyridine ≥97%
2-Fluoro-3-iodo-4-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅FINO
MW: 253.013 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28796391
CAS Number: 1627195-34-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR