2-Methyl-5-(methylthio)thiophene ≥97%

Supplier: AOBChem USA

Warning

57358-500MG 57358-1G 57358-5G 57358-25G
77329-916EA 214.24 USD
77329-916 77353-868 77366-228 77387-974
2-Methyl-5-(methylthio)thiophene ≥97%
2-Methyl-5-(methylthio)thiophene

Precaución: for Research Use Only

Formula: C₆H₈S₂
MW: 144.258 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11617751
CAS Number: 40990-29-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR