tert-Butyl 3,5-dimethyl-1H-pyrazole-1-carboxylate ≥97%

Supplier: AOBChem USA

Warning

57336-500MG 57336-1G 57336-5G 57336-25G
77329-912EA 122.57 USD
77329-912 77343-946 77366-224 77378-106
tert-Butyl 3,5-dimethyl-1H-pyrazole-1-carboxylate ≥97%
tert-Butyl 3,5-dimethyl-1H-pyrazole-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₆N₂O₂
MW: 196.246 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24474064
CAS Number: 219580-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR