1-Isobutyl-3,5-dimethyl-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

57335-500MG 57335-1G 57335-5G 57335-25G
77329-910EA 162.74 USD
77329-910 77346-074 77366-222 77387-970
1-Isobutyl-3,5-dimethyl-1H-pyrazole ≥97%
1-Isobutyl-3,5-dimethyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₉H₁₆N₂
MW: 152.237 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08701136
CAS Number: 79168-92-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR