1-(1-Ethoxyethyl)-3,5-dimethyl-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

57334-500MG 57334-1G 57334-5G 57334-25G
77329-908EA 159.65 USD
77329-908 77345-752 77366-220 77387-968
1-(1-Ethoxyethyl)-3,5-dimethyl-1H-pyrazole ≥97%
1-(1-Ethoxyethyl)-3,5-dimethyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₉H₁₆N₂O
MW: 168.236 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28168273
CAS Number: 1823538-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR