1-Ethyl-3,5-dimethyl-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

57332-1G 57332-5G 57332-25G
77341-286EA 81.37 USD
77341-286 77366-216 77378-102
1-Ethyl-3,5-dimethyl-1H-pyrazole ≥97%
1-Ethyl-3,5-dimethyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₇H₁₂N₂
Storage Temperature: Ambient
MDL Number: MFCD02601834
CAS Number: 17629-26-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR