4-Bromo-1-(2-methoxyethyl)-3,5-dimethyl-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

57330-500MG 57330-1G 57330-5G 57330-25G
77329-904EA 201.88 USD
77329-904 77353-866 77366-212 77387-966
4-Bromo-1-(2-methoxyethyl)-3,5-dimethyl-1H-pyrazole ≥97%
4-Bromo-1-(2-methoxyethyl)-3,5-dimethyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₈H₁₃BrN₂O
MW: 233.106 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16687003
CAS Number: 1342761-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR