4-Bromo-1-(methoxymethyl)-3,5-dimethyl-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

57329-500MG 57329-1G 57329-5G
77335-926EA 368.74 USD
77335-926 77353-864 77378-098
4-Bromo-1-(methoxymethyl)-3,5-dimethyl-1H-pyrazole ≥97%
4-Bromo-1-(methoxymethyl)-3,5-dimethyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₇H₁₁BrN₂O
MW: 219.079 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18324055
CAS Number: 1263210-45-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR