1-(4-Bromo-3,5-dimethyl-1H-pyrazol-1-yl)-2-methylpropan-2-ol ≥97%

Supplier: AOBChem USA

Warning

57328-500MG 57328-1G 57328-5G 57328-25G
77329-902EA 229.69 USD
77329-902 77353-862 77366-210 77387-964
1-(4-Bromo-3,5-dimethyl-1H-pyrazol-1-yl)-2-methylpropan-2-ol ≥97%
1-(4-Bromo-3,5-dimethyl-1H-pyrazol-1-yl)-2-methylpropan-2-ol

Precaución: for Research Use Only

Formula: C₉H₁₅BrN₂O
MW: 247.132 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16686976
CAS Number: 1250342-39-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR