4-Bromo-3,5-dimethyl-1-propyl-1H-pyrazole ≥97%
Supplier: AOBChem USA
|
Warning
|
57314-500MG
57314-1G
57314-5G
57314-25G
77329-900EA
234.84
USD
77329-900
77353-860
77366-206
77387-962
4-Bromo-3,5-dimethyl-1-propyl-1H-pyrazole ≥97%
4-Bromo-3,5-dimethyl-1-propyl-1H-pyrazole
Precaución: for Research Use Only
Formula:
C₈H₁₃BrN₂ MW: 217.106 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16687034 CAS Number: 1205195-05-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...