3-Bromo-5-iodo-4-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

57296-500MG 57296-1G 57296-5G 57296-25G
77329-898EA 86.52 USD
77329-898 77342-252 77366-198 77378-088
3-Bromo-5-iodo-4-methylbenzoic acid ≥97%
3-Bromo-5-iodo-4-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆BrIO₂
MW: 340.941 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24114741
CAS Number: 1229245-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR