1-Benzyl-4-bromo-3,5-dimethyl-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

57264-1G 57264-5G 57264-25G
77341-684EA 99.91 USD
77341-684 77366-192 77378-084
1-Benzyl-4-bromo-3,5-dimethyl-1H-pyrazole ≥97%
1-Benzyl-4-bromo-3,5-dimethyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₁₂H₁₃BrN₂
MW: 265.149 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00839492
CAS Number: 51108-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR