(2-Fluoro-4-iodo-3-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

57228-500MG 57228-1G 57228-5G 57228-25G
77329-892EA 126.69 USD
77329-892 77344-150 77366-186 77378-080
(2-Fluoro-4-iodo-3-methylphenyl)(methyl)sulfane ≥97%
(2-Fluoro-4-iodo-3-methylphenyl)(methyl)sulphane

Precaución: for Research Use Only

Formula: C₈H₈FIS
MW: 282.117 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758129
CAS Number: 2384707-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR