3-Fluoro-2-methyl-4-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

57224-500MG 57224-1G 57224-5G 57224-25G
77329-888EA 128.75 USD
77329-888 77344-258 77366-182 77378-078
3-Fluoro-2-methyl-4-(methylthio)benzoic acid ≥97%
3-Fluoro-2-methyl-4-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₂S
Storage Temperature: Ambient
MDL Number: MFCD32264106
CAS Number: 118287-06-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR