1,3-Dibromo-2-ethoxy-5-(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

57200-500MG 57200-1G 57200-5G 57200-25G
77329-884EA 146.26 USD
77329-884 77345-170 77366-176 77387-954
1,3-Dibromo-2-ethoxy-5-(trifluoromethyl)benzene ≥95%
1,3-Dibromo-2-ethoxy-5-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₉H₇Br₂F₃O
Storage Temperature: Ambient
MDL Number: MFCD34564436
CAS Number: 2384935-68-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR