2-(Benzyloxy)-1,3-dibromo-5-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

57199-500MG 57199-1G 57199-5G 57199-25G
77329-882EA 202.91 USD
77329-882 77353-852 77366-174 77387-952
2-(Benzyloxy)-1,3-dibromo-5-(trifluoromethyl)benzene ≥97%
2-(Benzyloxy)-1,3-dibromo-5-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₁₄H₉Br₂F₃O
Storage Temperature: Ambient
MDL Number: MFCD34530584
CAS Number: 2734774-20-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR