1,3-Dibromo-2-(methoxymethoxy)-5-(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

57197-500MG 57197-1G 57197-5G 57197-25G
77329-880EA 199.82 USD
77329-880 77353-850 77366-172 77387-950
1,3-Dibromo-2-(methoxymethoxy)-5-(trifluoromethyl)benzene ≥95%
1,3-Dibromo-2-(methoxymethoxy)-5-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₉H₇Br₂F₃O₂
MW: 363.954 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530582
CAS Number: 2734773-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR