2-Bromo-3,4-dichlorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

57190-500MG 57190-1G 57190-5G
77339-544EA 596.37 USD
77339-544 77353-846 77378-068
2-Bromo-3,4-dichlorobenzaldehyde ≥97%
2-Bromo-3,4-dichlorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃BrCl₂O
Storage Temperature: Ambient
MDL Number: MFCD34758106
CAS Number: 2091746-50-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR