(2-Bromo-4,5-dichlorophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

57189-500MG 57189-1G 57189-5G
77337-446EA 414.06 USD
77337-446 77353-844 77378-066
(2-Bromo-4,5-dichlorophenyl)methanol ≥97%
(2-Bromo-4,5-dichlorophenyl)methanol

Precaución: for Research Use Only

Formula: C₇H₅BrCl₂O
MW: 255.924 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28737833
CAS Number: 1067882-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR