4,5-Dichloro-2-iodobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

57187-500MG 57187-1G 57187-5G
77340-100EA 754.99 USD
77340-100 77353-842 77378-064
4,5-Dichloro-2-iodobenzaldehyde ≥97%
4,5-Dichloro-2-iodobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃Cl₂IO
MW: 300.909 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758104
CAS Number: 2092867-38-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR